ENA333558937 Enamine

4-(3-chlorophenoxymethyl)benzoic acid

  • MDL number MFCD02090855
  • Cas Number : 149288-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83485

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83485
Availability details for
Quantity
  • Availability
  • Add to Cart