ENA333558946 Enamine

3-(2-chlorophenoxymethyl)benzoic acid

  • MDL number MFCD00625985
  • Cas Number : 30082-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83494

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83494
Availability details for
Quantity
  • Availability
  • Add to Cart