ENA333558947 Enamine

5-[(3-methoxyphenoxy)methyl]furan-2-carboxylic acid

  • MDL number MFCD02090866
  • Cas Number : 405897-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83495
Availability details for
Quantity
  • Availability
  • Add to Cart