ENA333558948 Enamine

5-(2,5-dimethylphenoxymethyl)furan-2-carboxylic acid

  • MDL number MFCD02090870
  • Cas Number : 364736-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83496

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83496
Availability details for
Quantity
  • Availability
  • Add to Cart