ENA333558954 Enamine

5-(2,5-dichlorophenoxymethyl)furan-2-carboxylic acid

  • MDL number MFCD01858840
  • Cas Number : 364616-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83502

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83502
Availability details for
Quantity
  • Availability
  • Add to Cart