ENA333558955 Enamine

5-(2-chlorophenoxymethyl)furan-2-carboxylic acid

  • MDL number MFCD01855739
  • Cas Number : 364609-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83503

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83503
Availability details for
Quantity
  • Availability
  • Add to Cart