ENA333558960 Enamine

3-[(1H-pyrazol-1-yl)methyl]benzoic acid

  • MDL number MFCD02253844
  • Cas Number : 562803-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83508
Availability details for
Quantity
  • Availability
  • Add to Cart