ENA333558966 Enamine

3-(3-chlorophenoxymethyl)benzoic acid

  • MDL number MFCD02257635
  • Cas Number : 360778-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83514

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83514
Availability details for
Quantity
  • Availability
  • Add to Cart