ENA333558967 Enamine

5-[(4-nitro-1H-pyrazol-1-yl)methyl]furan-2-carboxylic acid

  • MDL number MFCD02055758
  • Cas Number : 436086-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83515

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83515
Availability details for
Quantity
  • Availability
  • Add to Cart