ENA333558969 Enamine

5-(4-chlorophenoxymethyl)furan-2-carboxylic acid

  • MDL number MFCD02253883
  • Cas Number : 74556-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83517

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83517
Availability details for
Quantity
  • Availability
  • Add to Cart