ENA333558970 Enamine

5-(4-ethylphenoxymethyl)furan-2-carboxylic acid

  • MDL number MFCD02253884
  • Cas Number : 364595-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83518

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83518
Availability details for
Quantity
  • Availability
  • Add to Cart