ENA333558971 Enamine

4-(2-chlorophenoxymethyl)benzoic acid

  • MDL number MFCD01893322
  • Cas Number : 149288-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83519

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83519
Availability details for
Quantity
  • Availability
  • Add to Cart