ENA333558972 Enamine

4-[(4-chloro-3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzoic acid

  • MDL number MFCD01853371
  • Cas Number : 376615-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83520

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83520
Availability details for
Quantity
  • Availability
  • Add to Cart