ENA333558983 Enamine

3-[(2,2,3,3-tetrafluoropropoxy)methyl]benzoic acid

  • MDL number MFCD02253707
  • Cas Number : 438466-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83531

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83531
Availability details for
Quantity
  • Availability
  • Add to Cart