ENA333558985 Enamine

5-(3,5-dimethylphenoxymethyl)furan-2-carboxylic acid

  • MDL number MFCD02253718
  • Cas Number : 405898-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83533

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83533
Availability details for
Quantity
  • Availability
  • Add to Cart