ENA333558990 Enamine

4-iodo-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD00465280
  • Cas Number : 6647-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 233...235
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83538

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-83538
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart