ENA333558991 Enamine

2-(3-methoxy-4-nitro-1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD03074438
  • Cas Number : 512809-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83540
Availability details for
Quantity
  • Availability
  • Add to Cart