ENA333558995 Enamine

3,5-bis(methylsulfanyl)-1,2-thiazole-4-carboxylic acid

  • MDL number MFCD00737783
  • Cas Number : 4886-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83544

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83544
Availability details for
Quantity
  • Availability
  • Add to Cart