ENA333559001 Enamine

1-(4-nitrophenyl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD00541956
  • Cas Number : 82552-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83551
Availability details for
Quantity
  • Availability
  • Add to Cart