ENA333559004 Enamine

2-[3-(1H-1,2,3,4-tetrazol-1-yl)phenoxy]propanoic acid

  • MDL number MFCD03419508
  • Cas Number : 832739-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83554
Availability details for
Quantity
  • Availability
  • Add to Cart