ENA333559005 Enamine

2-(5-amino-2H-1,2,3,4-tetrazol-2-yl)acetic acid

  • MDL number MFCD03419553
  • Cas Number : 21743-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83555

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83555
Availability details for
Quantity
  • Availability
  • Add to Cart