ENA333559007 Enamine

2-(3-methyl-4-nitro-1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD04967123
  • Cas Number : 956204-54-9
  • Molecular Weight : 185.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-83557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83557
Availability details for
Quantity
  • Availability
  • Add to Cart