ENA333559009 Enamine

2-(4-chloro-5-methyl-1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD03419600
  • Cas Number : 180741-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83559
Availability details for
Quantity
  • Availability
  • Add to Cart