ENA333559013 Enamine

3-(4-bromo-3-methyl-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD03419613
  • Cas Number : 956824-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83563

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83563
Availability details for
Quantity
  • Availability
  • Add to Cart