ENA333559017 Enamine

3-(4-chloro-5-methyl-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD03419616
  • Cas Number : 180741-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83567
Availability details for
Quantity
  • Availability
  • Add to Cart