ENA333559019 Enamine

3-(5-methyl-4-nitro-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD03419618
  • Cas Number : 1004644-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83569
Availability details for
Quantity
  • Availability
  • Add to Cart