ENA333559024 Enamine

2-(3-methyl-4-nitro-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD03419633
  • Cas Number : 956950-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83574

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83574
Availability details for
Quantity
  • Availability
  • Add to Cart