ENA333559031 Enamine

3-(3-nitro-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD03419659
  • Cas Number : 1004644-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83581

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83581
Availability details for
Quantity
  • Availability
  • Add to Cart