ENA333559034 Enamine

3-(4-chloro-3,5-dimethyl-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD03419665
  • Cas Number : 890597-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-83584

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83584
Availability details for
Quantity
  • Availability
  • Add to Cart