ENA333559035 Enamine

3-(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD03074433
  • Cas Number : 512809-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83585

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83585
Availability details for
Quantity
  • Availability
  • Add to Cart