ENA333559044 Enamine

2-[4-bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

  • MDL number MFCD03419689
  • Cas Number : 1005584-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83594
Availability details for
Quantity
  • Availability
  • Add to Cart