ENA333559045 Enamine

3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

  • MDL number MFCD03419693
  • Cas Number : 925606-77-5
  • Molecular Weight : 222.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-83595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83595
Availability details for
Quantity
  • Availability
  • Add to Cart