ENA333559052 Enamine

2-[4-chloro-3,5-bis(difluoromethyl)-1H-pyrazol-1-yl]acetic acid

  • MDL number MFCD03419712
  • Cas Number : 1005631-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83602

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83602
Availability details for
Quantity
  • Availability
  • Add to Cart