ENA333559053 Enamine

2-[3,5-bis(difluoromethyl)-1H-pyrazol-1-yl]propanoic acid

  • MDL number MFCD03419715
  • Cas Number : 1005586-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-83603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83603
Availability details for
Quantity
  • Availability
  • Add to Cart