ENA333559054 Enamine

2-[4-chloro-3,5-bis(difluoromethyl)-1H-pyrazol-1-yl]propanoic acid

  • MDL number MFCD03419716
  • Cas Number : 1005678-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83604
Availability details for
Quantity
  • Availability
  • Add to Cart