ENA333559056 Enamine

2-[4-chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid

  • MDL number MFCD03419740
  • Cas Number : 1006447-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83606

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83606
Availability details for
Quantity
  • Availability
  • Add to Cart