ENA333559062 Enamine

3-[4-chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

  • MDL number MFCD03419752
  • Cas Number : 1001518-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83612

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83612
Availability details for
Quantity
  • Availability
  • Add to Cart