ENA333559065 Enamine

4-chloro-1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD03419816
  • Cas Number : 1001519-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83615

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83615
Availability details for
Quantity
  • Availability
  • Add to Cart