ENA333559071 Enamine

5-bromo-2-(difluoromethoxy)benzoic acid

  • MDL number MFCD03420217
  • Cas Number : 438221-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83621

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83621
Availability details for
Quantity
  • Availability
  • Add to Cart