ENA333559074 Enamine

5-(3-chloro-4-fluorophenoxymethyl)furan-2-carboxylic acid

  • MDL number MFCD03421870
  • Cas Number : 832737-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83624

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83624
Availability details for
Quantity
  • Availability
  • Add to Cart