ENA333559077 Enamine

5-(2-methylphenoxymethyl)furan-2-carboxylic acid

  • MDL number MFCD03421877
  • Cas Number : 832739-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83627

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83627
Availability details for
Quantity
  • Availability
  • Add to Cart