ENA333559082 Enamine

4-bromo-1-ethyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD03074558
  • Cas Number : 512810-22-9
  • Molecular Weight : 219.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-83632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83632
Availability details for
Quantity
  • Availability
  • Add to Cart