ENA333559087 Enamine

4-methyl-5-phenylthiophene-2-carboxylic acid

  • MDL number MFCD01830424
  • Cas Number : 40133-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83637

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83637
Availability details for
Quantity
  • Availability
  • Add to Cart