ENA333559088 Enamine

5,7-bis(difluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD03466608
  • Cas Number : 438227-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83638
Availability details for
Quantity
  • Availability
  • Add to Cart