ENA333559098 Enamine

3-(3,5-dimethyl-4-nitro-1H-pyrazol-1-yl)-2-methylpropanoic acid

  • MDL number MFCD04967183
  • Cas Number : 1005640-74-3
  • Molecular Weight : 227.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-83648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83648
Availability details for
Quantity
  • Availability
  • Add to Cart