ENA333559100 Enamine

5-cyclopropyl-7-(difluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD04967184
  • Cas Number : 832739-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83650
Availability details for
Quantity
  • Availability
  • Add to Cart