ENA333559101 Enamine

3-(3-methyl-2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoic acid

  • MDL number MFCD04967185
  • Cas Number : 832740-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83651

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83651
Availability details for
Quantity
  • Availability
  • Add to Cart