ENA333559103 Enamine

7-(difluoromethyl)-5-methyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD04967190
  • Cas Number : 725699-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83653
Availability details for
Quantity
  • Availability
  • Add to Cart