ENA333559104 Enamine

7-(chlorodifluoromethyl)-5-methylpyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD04045223
  • Cas Number : 676522-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83654
Availability details for
Quantity
  • Availability
  • Add to Cart