ENA333559105 Enamine

2-[4-chloro-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid

  • MDL number MFCD04967193
  • Cas Number : 1002032-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83655
Availability details for
Quantity
  • Availability
  • Add to Cart